missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TGM2 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human TGM2 (NP_004604.2) (Met 1-Ala 687) fused to a polyhistidine tag at the N-terminus

$210.00 - $359.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias RP5-1054A22.2, G-ALPHA-h, GNAH, TG2, TGC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11095H07B25 View Documents Life Technologies
LS11095H07B25
5 x 5μg Each for $210.00
 
11095H07B50 View Documents Life Technologies
LS11095H07B50
50μg Each for $359.00
 
Description

Description

Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase) (TGM2), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP5-1054A22.2, G-ALPHA-h, GNAH, TG2, TGC
PAGE
Frozen
7052

For Research Use Only. Not for use in diagnostic procedures.