Please login to your online account to display your discounted pricing

Sino Biological™ TGM2 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human TGM2 (NP_004604.2) (Met 1-Ala 687) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11095H07B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11095H07B25

  • / Each
 

Description & Specifications

Specifications

Gene Symbol TGM2
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Frozen
Purity ≥90% by SDS-PAGE
Purification Method PAGE
Gene Alias RP5-1054A22.2, G-ALPHA-h, GNAH, TG2, TGC
Expression System Insect
Endotoxin Concentration <1 EU/μg
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Validated Application Assay Standard
Quantity 5 x 5μg
Molecular Weight 79.6kDa
Structural Form Full Length/Recombinant
Common Name Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase)
Gene ID (Entrez) 7052

Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase) (TGM2), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.