Please login to your online account to display your discounted pricing

Sino Biological™ TFRC Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TFRC (NP_003225.2) (Cys 89-Phe 760) fused to a polyhistidine tag at the N-terminus

$90.50 - $1,294.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD71, TFR, TFR1, TRFR
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11020H07H25

DFS Item
 
life technologies
LS11020H07H25
5 x 5μg Each for $206.00

11020H07H5

DFS Item
 
life technologies
LS11020H07H5
5μg Each for $90.50

11020H07H50

DFS Item
 
life technologies
LS11020H07H50
50μg Each for $357.00

11020H07HL

DFS Item
 
life technologies
LS11020H07H250
5 x 50μg Each for $1,294.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD71, TFR, TFR1, TRFR

Transferrin Receptor (p90 CD71) (TFRC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TFRC at 10μg/mL (100μL/well) can bind human Transferrin, the EC50 of human Transferrin is 5.6ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.