missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TACSTD2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) fused to a polyhistidine tag at the C-terminus

$278.00 - $278.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TACSTD2, EGP-1, GA733, GA733-1, M1S1, TROP2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10428H08H50 View Documents Life Technologies
LS10428H08H50
50μg Each for $278.00
 
Description

Description

Tumor-Associated Calcium Signal Transducer 2 (TACSTD2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
TACSTD2, EGP-1, GA733, GA733-1, M1S1, TROP2
PAGE
Lyophilized
4070

For Research Use Only. Not for use in diagnostic procedures.