Please login to your online account to display your discounted pricing

Sino Biological™ TACSTD2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10428H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10428H08H50

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Structural Form Truncated/Recombinant
Expression System Mammalian
Validated Application Assay Standard
Gene ID (Entrez) 4070
Molecular Weight 29.4kDa
Common Name Tumor-associated calcium signal transducer 2
Protein Family Other Proteins
Protein Tag His Tag
Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Quantity 50μg
Purification Method PAGE
Termination C-Terminal
Gene Symbol TACSTD2
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias TACSTD2, EGP-1, GA733, GA733-1, M1S1, TROP2

Tumor-Associated Calcium Signal Transducer 2 (TACSTD2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.