missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SUMO1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SUMO1 (AAH66306.1) (Ser 2-Val 101) fused to a polyhistidine tag at the N-terminus

$752.00 - $752.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias OK/SW-cl.43, DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
13095H07E50 View Documents Life Technologies
LS13095H07E50
50μg Each for $752.00
 
Description

Description

SMT3 Suppressor of Mif Two 3 Homolog (SUMO1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
OK/SW-cl.43, DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1
PAGE
Lyophilized
7341

For Research Use Only. Not for use in diagnostic procedures.