Please login to your online account to display your discounted pricing

Sino Biological™ SPINT1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SPINT1 (O43278-2) (Met 1-Val 433) fused to a polyhistidine tag at the C-terminus

$228.00 - $660.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SPINT1, UNQ223/PRO256, HAI, HAI1, MANSC2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11742H08H25

DFS Item
 
life technologies
LS11742H08H25
5 x 5μg Each for $660.00

11742H08H5

DFS Item
 
life technologies
LS11742H08H5
5μg Each for $228.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SPINT1, UNQ223/PRO256, HAI, HAI1, MANSC2

Serine Peptidase Inhibitor Kazal Type 1 (SPINT1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (R&D Systems Cat. No. ES002); the IC50 value is <2nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.