Please login to your online account to display your discounted pricing

Sino Biological™ SPINT1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human SPINT1 (O43278-2) (Met 1-Val 433) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11742H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11742H08H25

  • / Each
 

Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias SPINT1, UNQ223/PRO256, HAI, HAI1, MANSC2
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Endotoxin Concentration <1 EU/μg
Quantity 5 x 5μg
Protein Subtype Protease Inhibitors
Protein Family Protease Inhibitors
Molecular Weight 45.8kDa
Gene Symbol SPINT1
Expression System Mammalian
Format Lyophilized
Validated Application Assay Standard
Common Name Serine peptidase inhibitor, Kazal type 1
Protein Tag His Tag

Serine Peptidase Inhibitor Kazal Type 1 (SPINT1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (R&D Systems Cat. No. ES002); the IC50 value is <2nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.