Please login to your online account to display your discounted pricing

Sino Biological™ SPARC Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human SPARC (NP_003109.1) precursor (Met 1-Ile 303) fused to a carboxy-terminal polyhistidine tag

Manufacturer: life technologies  LS10929H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10929H08H50

  • / Each
 

Description & Specifications

Specifications

Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 34kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Common Name Secreted protein, acidic, cysteine-rich glycoprotein
Gene ID (Entrez) 6678
Format Lyophilized
Structural Form Full Length/Recombinant
Purification Method PAGE
Termination C-Terminal
Quantity 50μg
Validated Application Assay Standard
Gene Alias ON, SPARC
Protein Tag His Tag
Gene Symbol SPARC
Protein Family Other Proteins
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg

Secreted Protein, Acidic, Cysteine-Rich Glycoprotein (SPARC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.