Please login to your online account to display your discounted pricing

Sino Biological™ SOST Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human SOST (NP_079513.1) (Gln 24-Tyr 213) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10593H07H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10593H07H50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Purification Method PAGE
Validated Application Assay Standard
Purity ≥85% by SDS-PAGE
Format Lyophilized
Termination N-Terminal
Structural Form Full Length/Recombinant
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 22.5kDa
Gene Alias Sclerosteosis, SOST, VBCH
Gene Symbol SOST
Gene ID (Entrez) 50964
Expression System Mammalian
Common Name Sclerostin
Protein Tag His Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration <1 EU/μg
Protein Family Other Proteins

Sclerostin (SOST), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.