missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ SLPI Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the secreted form of human SLPI (P03973) (Ser 26-Ala 132) fused to a polyhistidine tag at the N-terminus

$78.00 - $229.00

Specifications

Purity >84% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10343H07B25 Life Technologies
LS10343H07B25
5 x 5μg Each for $172.00
10343H07B5 Life Technologies
LS10343H07B5
5μg Each for $78.00
10343H07B50 Life Technologies
LS10343H07B50
50μg Each for $229.00
Specifications

Description

Secretary Leukocyte Peptidase Inhibitor (SLPI), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>84% by SDS-PAGE
<1 EU/μg
ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
PAGE
Frozen
6590