Please login to your online account to display your discounted pricing

Sino Biological™ SLPI Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the secreted form of human SLPI (P03973) (Ser 26-Ala 132) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10343H07B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10343H07B5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 14kDa
Purification Method PAGE
Purity >84% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
Common Name Secretory Leukocyte Peptidase Inhibitor
Gene Symbol SLPI
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene ID (Entrez) 6590

Secretary Leukocyte Peptidase Inhibitor (SLPI), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.