missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ SLAMF1 (CD150) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SLAMF1 precursor (NP_003028.1), residues 1 to 258, fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10837H08H250

 View more versions of this product

Catalog No. 10837H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; SLAMF1 (CD150) Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Signaling lymphocytic activation molecule (SLAMF1/CD150), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human SLAMF1-His at 1μg/well can bind biotinylated recombinant human SH2D1A with a linear range of 0.8-20.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6504
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD150, CDw150, SLAM, SLAMF1
SLAMF1
CD Proteins
His Tag
Lyophilized
25.8kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Signaling lymphocytic activation molecule family member 1
Assay Standard
Mammalian