missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SIRPA Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SIRPA (NP_542970.1) extracellular domain (Met 1-Arg 370) fused to a polyhistidine tag at the C-terminus

$102.00 - $102.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP, SIRPA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11612H08H5 DFS Item
View Documents
Invitrogen
LS11612H08H5
5μg
Each for $102.00
 
Description

Description

Signal-Regulatory Protein Alpha (SIRPA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP, SIRPA
PAGE
Lyophilized
140885
SDS

For Research Use Only. Not for use in diagnostic procedures.