Please login to your online account to display your discounted pricing

Sino Biological™ SERPINE1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human SerpinE1 precursor (NP_000593.1) (Met 1-Pro 402) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10296H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10296H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 44.2kDa
Purification Method PAGE
Purity ≥95% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias SERPINE1, PAI, PAI-1, PAI1, PLANH1
Common Name Serpin peptidase inhibitor, clade E, member 1
Gene Symbol SERPINE1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 5054

Serpin Peptidase Inhibitor, Clade E, Member 1 (SERPINE1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM NaAc, 100mM NaCl, pH5.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.