missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SERPINA3 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SERPINA3 (NP_001076.2) (Met 1-Ala 423) fused to a C-terminal polyhistidine tag

$231.00 - $231.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SERPINA3, AACT, ACT, GIG24, GIG25, mgC88254
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10307H08H5 View Documents Life Technologies
LS10307H08H5
5μg Each for $231.00
 
Description

Description

Serine Peptidase Inhibitor Clade A Member 3 (SERPINA3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Anaspec Cat. No. 27114), the IC50 value is <20nM
  • Formulation: Lyophilized in 25mM HEPES, 0.15M NaCl, pH 7.8, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
SERPINA3, AACT, ACT, GIG24, GIG25, mgC88254

For Research Use Only. Not for use in diagnostic procedures.