Please login to your online account to display your discounted pricing

Sino Biological™ SERPINA3 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human SERPINA3 (NP_001076.2) (Met 1-Ala 423) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10307H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10307H08H5

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Protein Tag His Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5μg
Purity ≥90% by SDS-PAGE
Gene Symbol SERPINA3
Protein Family Protease Inhibitors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Expression System Mammalian
Molecular Weight 46.5kDa
Protein Subtype Proteinase Inhibitor
Endotoxin Concentration <1 EU/μg
Gene Alias SERPINA3, AACT, ACT, GIG24, GIG25, mgC88254
Common Name Serine peptidase inhibitor, clade A, member 3

Serine Peptidase Inhibitor Clade A Member 3 (SERPINA3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Anaspec Cat. No. 27114), the IC50 value is <20nM
  • Formulation: Lyophilized in 25mM HEPES, 0.15M NaCl, pH 7.8, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.