missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SERPINA1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SERPINA1 pre-protein (NP_000286.3) (Met 1-Lys 418) fused to the polyhistidine tag at the C-terminus

$245.00 - $722.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SerpinA1, PI, A1A, AAT, PI1, A1AT, mgC9222, PRO2275, mgC23330
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10306H08H25 View Documents Life Technologies
LS10306H08H25
5 x 5μg Each for $722.00
 
10306H08H5 View Documents Life Technologies
LS10306H08H5
5μg Each for $245.00
 
Description

Description

Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1 (SERPINA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2
    ; the IC50 value is <3.0nM, as measured in 100μL reaction mixture containing 1.25ng trypsin, 10μM substrate, 50mM Tris, 10mM CaCl2, 0.15 M NaCl, pH 7.5
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
SerpinA1, PI, A1A, AAT, PI1, A1AT, mgC9222, PRO2275, mgC23330

For Research Use Only. Not for use in diagnostic procedures.