missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ SEMA5A Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SEMA5A (NP_003957.2) extracellular domain (Met 1-Met 968) fused to a polyhistidine tag at the C-terminus

$634.00 - $634.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FLJ12815, SEMAF, semF, SEMA5A
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11300H08H50 Life Technologies
LS11300H08H50
50μg Each for $634.00
Specifications

Description

Sema Domain, Seven Thrombospondin Repeats (TM) and Short Cytoplasmic Domain, (Semaphorin) 5A (SEMA5A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
FLJ12815, SEMAF, semF, SEMA5A
PAGE
Lyophilized
9037