missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ SEMA5A Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SEMA5A (NP_003957.2) extracellular domain (Met 1-Met 968) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11300H08H50

 View more versions of this product

Catalog No. 11300H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; SEMA5A Recombinant Human Protein, His Tag; 50ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Sema Domain, Seven Thrombospondin Repeats (TM) and Short Cytoplasmic Domain, (Semaphorin) 5A (SEMA5A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9037
PAGE
50μg
C-Terminal
<1 EU/μg
Sema domain, 7 thrombospondin repeats, TM and short cytoplasmic domain, (semaphorin) 5A
Assay Standard
Mammalian
Lyophilized
107kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
FLJ12815, SEMAF, semF, SEMA5A
SEMA5A
Other Proteins
His Tag