Please login to your online account to display your discounted pricing

Sino Biological™ SEMA4D Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse SEMA4D (NP_038688.2) extracellular domain (Met 1-Arg 733) fused to a C-terminal polyhistidine tag

$322.00 - $1,150.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD100, Semacl2, Semaj, Semcl2, coll-4
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50642M08H50

DFS Item
 
life technologies
LS50642M08H50
50μg Each for $322.00

50642M08HL

DFS Item
 
life technologies
LS50642M08H250
5 x 50μg Each for $1,150.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD100, Semacl2, Semaj, Semcl2, coll-4

Sema Domain Immunoglobulin Domain (Ig), Tm and Short Cytoplasmic Domain, (Semaphorin) 4D (SEMA4D), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind human SEMA4A in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.