Please login to your online account to display your discounted pricing

Sino Biological™ SEMA4A Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human SEMA4A (NP_071762.2) (Met 1-His 683) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11756H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11756H08HL

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 73.4kDa
Protein Subtype Other Proteins
Protein Family Other Proteins
Purity ≥90% by SDS-PAGE
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Expression System Mammalian
Common Name Sema domain, immunoglobulin domain (Ig), TM and short cytoplasmic domain, (semaphorin) 4A
Validated Application Assay Standard
Gene Symbol SEMA4A
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5 x 50μg
Gene Alias RP11-54H19.2, CORD10, FLJ12287, RP35, SEMAB, SEMB

Sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4 (SEMA4A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated mouse SEMA4D in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.