missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SEMA3A Recombinant Mouse Protein, N-hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Lys 26-Phe 546) of mouse SEMA3A (O08665) fused to the Fc region of human IgG1 at the N-terminus

$264.00 - $510.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Hsema-I, SEMA1, SemD, Semad, coll-1
View More Specs

Includes: Five vials of protein in suspension buffer

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50631M01H25 View Documents Life Technologies
LS50631M01H25
5 x 5μg Each for $264.00
 
50631M01H50 View Documents Life Technologies
LS50631M01H50
50μg Each for $510.00
 
Description

Description

Sema Domain, Immunoglobulin Domain (Ig) Short Basic Domain, Secreted, (Semaphorin) 3A (SEMA3A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Hsema-I, SEMA1, SemD, Semad, coll-1
PAGE
Lyophilized
20346

For Research Use Only. Not for use in diagnostic procedures.