missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SELPLG (PSGL1) Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 295) of human PSGL-1 precursor (NP_002997.1) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$102.00 - $381.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SELPLG, CD162, CLA, PSGL-1, PSGL1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10490H03H5 View Documents Life Technologies
LS10490H03H5
5μg Each for $102.00
 
10490H03H50 View Documents Life Technologies
LS10490H03H50
50μg Each for $381.00
 
Description

Description

Selectin P Ligand (SELPLG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
SELPLG, CD162, CLA, PSGL-1, PSGL1
PAGE
Lyophilized
6404

For Research Use Only. Not for use in diagnostic procedures.