Please login to your online account to display your discounted pricing

Sino Biological™ SELE (E-SELECTIN) Recombinant Rat Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the rat SELE (P98105-1) extracellular domain (Met 1-Pro 494) fused to a polyhistidine tag at the C-terminus

$92.75 - $638.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SELE
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

80033R08H25

DFS Item
 
life technologies
LS80033R08H25
5 x 5μg Each for $200.00

80033R08H5

DFS Item
 
life technologies
LS80033R08H5
5μg Each for $92.75

80033R08HL

DFS Item
 
life technologies
LS80033R08H250
5 x 50μg Each for $638.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SELE

Selectin E (SELE), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when 5 x 104 cells/well are added to rat SELE coated plates (2μg/mL with 100μL/well), approximately 20-35% will adhere for 1 hour incubation at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.