Please login to your online account to display your discounted pricing

Sino Biological™ SELE (E-SELECTIN) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 556) of the human SELE precursor (NP_000441.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10335H03H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10335H03H5

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Molecular Weight 86.5kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias SELE, RP1-117P20.2, CD62E, ELAM, ELAM1, ESEL, LECAM2
Common Name SELE selectin E
Gene Symbol SELE
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
Protein Tag His Tag/IgG1 Fc Tag

Selectin E (SELE) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.&line-break;Background: E-selectin, also known as endothelial leukocyte adhesion molecule-1 (ELAM-1) and CD62E, is an inducible adhesion molecule that is expressed at relatively high density on the surface of cultured endothelial cells that have been activated by proinflammatory cytokines, such as IL- 1β, TNF-α, or bacterial endotoxin. E-selectin exhibits a complex mosaic structure consisting of a large extracellular region comprised of a lectin domain, an EGF-like domain, and a short consensus repeat (SCR) domain, followed by a transmembrane region and a relatively short (32 aa) cytoplasmic tail. As a member of the LEC-CAM or selectin family, E-selectin recognises and binds to sialylated carbohydrates including members of the Lewis X and Lewis A families found on monocytes, granulocytes, and T-lymphocytes. E-selectin supports rolling and stable arrest of leukocytes on activated vascular endothelium, and furthermore, it was indicated that it can also transduce an activating stimulus via the MAPK cascade into the endothelial cell during leukocyte adhesion. In addition, this adhesion molecule appears to be involved in the pathogenesis of atherosclerosis.

  • N-terminal Sequence Analysis: Trp 22
  • Activity: measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when cells are added to SELE coated plates (2μg/mL, 100μL/well) approximately 90-100 % will adhere specifically
  • Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.