Please login to your online account to display your discounted pricing

Sino Biological™ SECTM1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human SECTM1 (Q8WVN6-1) extracellular domain (Met 1-Gly 145) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS12269H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12269H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 14kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias K12
Format Lyophilized
Purification Method PAGE
Expression System Mammalian
Quantity 50μg
Gene Symbol SECTM1
Common Name Secreted and transmembrane 1
Gene ID (Entrez) 6398
Protein Family Other Proteins
Protein Tag His Tag
Validated Application Assay Standard
Structural Form Truncated/Recombinant
Termination C-Terminal
Purity ≥90% by SDS-PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration <1 EU/μg

Secreted and Transmembrane 1 (SECTM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.