missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SCARB2 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human SCARB2 (NP_005497.1) extracellular domain (Arg 27-Thr 432) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$213.00 - $1,154.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias SCARB2, AMRF, CD36L2, HLGP85, LIMPII, SR-BII
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11063H03H25 View Documents Life Technologies
LS11063H03H25
5 x 5μg Each for $213.00
 
11063H03HL View Documents Life Technologies
LS11063H03H250
5 x 50μg Each for $1,154.00
 
Description

Description

Scavenger receptor class B, member 2 (SCARB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human RSPO1 in a functional ELISA
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
SCARB2, AMRF, CD36L2, HLGP85, LIMPII, SR-BII

For Research Use Only. Not for use in diagnostic procedures.