Please login to your online account to display your discounted pricing

Sino Biological™ S100B Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to a polyhistidine tag at the N-terminus

$201.00 - $1,122.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias NEF, S100, S100beta
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10181H07E25

DFS Item
 
life technologies
LS10181H07E25
5 x 5μg Each for $201.00

10181H07E50

DFS Item
 
life technologies
LS10181H07E50
50μg Each for $316.00

10181H07EL

DFS Item
 
life technologies
LS10181H07E250
5 x 50μg Each for $1,122.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias NEF, S100, S100beta

S100 Calcium Binding Protein B (S100B), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its ability to bind human AGER in a functional ELISA; 2. Measured by its ability to bind TP53 in a functional ELISA
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.