missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ S100B Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Life Technologies LS10181H07E250

 View more versions of this product

Catalog No. 10181H07EL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; S100B Recombinant Human Protein, N-His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

S100 Calcium Binding Protein B (S100B), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human AGER in a functional ELISA; 2. Measured by its ability to bind TP53 in a functional ELISA
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12kDa
5 x 50μg
Neurotrophic Factors|Other Proteins
S100 calcium binding protein B
Assay Standard
His Tag
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
NEF, S100, S100beta
S100B
E. coli