Please login to your online account to display your discounted pricing

Sino Biological™ S100B Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10181H01H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10181H01H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 37.2kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Quantity 50μg
Protein Subtype Neurotrophic Factors|Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias NEF, S100, S100beta
Common Name S100 calcium binding protein B
Gene Symbol S100B
Protein Tag IgG1 Fc Tag
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

S100 calcium binding protein B (S100B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a functional ELISA: 1. Measured by its ability to bind mouse S100A1; 2. Measured by its ability to bind TP53
  • Formulation: Lyophilized in 100mM Gly, 10mM NaCl, 50mM Tris, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.