Please login to your online account to display your discounted pricing

Sino Biological™ S100A9 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human S100A9 (NP_002956.1) (Met 1-Pro 114)

Manufacturer: life technologies  LS11145HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11145HNAE50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 26kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias 60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
Common Name S100 calcium binding protein A9
Gene Symbol S100A9
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

S100 Calcium Binding Protein A9 (S100A9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells; the ED50 for this effect is typically 3-15μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.