missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S100A8 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100A8 (NP_002955.2) (Leu 2-Glu 93) fused to a polyhistidine tag at the N-terminus

$110.00 - $355.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11138H07E5 View Documents Life Technologies
LS11138H07E5
5μg Each for $110.00
 
11138H07E50 View Documents Life Technologies
LS11138H07E50
50μg Each for $355.00
 
Description

Description

S100 Calcium Binding Protein A8 (S100A8), recombinant human protein is supplied in frozen format. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human S100A9 in a functional ELISA; measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells, the ED50 for this effect is typically 10-30μg/mL
  • Formulation: 20mM Tris, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8

For Research Use Only. Not for use in diagnostic procedures.