missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S100A6 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100A6 (NP_055439.1) (Met 1-Gly 90) fused to a C-terminal polyhistidine tag

$360.00 - $560.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 2A9, 5B10, CABP, CACY, PRA, S100A6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10939H08B25 View Documents Life Technologies
LS10939H08B25
5 x 5μg Each for $360.00
 
10939H08B50 View Documents Life Technologies
LS10939H08B50
50μg Each for $560.00
 
Description

Description

S100 Calcium Binding Protein A6 (S100A6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, 0.5mM PMSF, 1mM TCEP, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
2A9, 5B10, CABP, CACY, PRA, S100A6
PAGE
Lyophilized
6277

For Research Use Only. Not for use in diagnostic procedures.