Please login to your online account to display your discounted pricing

Sino Biological™ S100A6 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human S100A6 (NP_055439.1) (Met 1-Gly 90) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10939H08B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10939H08B25

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥95% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 12kDa
Validated Application Assay Standard
Termination C-Terminal
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Common Name S100 calcium binding protein A6
Gene ID (Entrez) 6277
Expression System Insect
Gene Symbol S100A6
Protein Family Other Proteins
Protein Subtype Other Proteins
Structural Form Full Length/Recombinant
Format Lyophilized
Purification Method PAGE
Gene Alias 2A9, 5B10, CABP, CACY, PRA, S100A6

S100 Calcium Binding Protein A6 (S100A6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, 0.5mM PMSF, 1mM TCEP, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

This protein is manufactured by Sino Biological Inc.