missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S100A4 Recombinant Human Protein, N-hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100A4 (NP_002952.1) (Met 1-Lys 101) fused to Fc region of human IgG1 at the N-terminus

$395.00 - $395.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10185H01H50 View Documents Life Technologies
LS10185H01H50
50μg Each for $395.00
 
Description

Description

S100 Calcium Binding Protein A4 (S100A4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98
PAGE
Lyophilized
6275

For Research Use Only. Not for use in diagnostic procedures.