Please login to your online account to display your discounted pricing

Sino Biological™ S100A4 Recombinant Human Protein, N-hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human S100A4 (NP_002952.1) (Met 1-Lys 101) fused to Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10185H01H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10185H01H50

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene ID (Entrez) 6275
Common Name S100 calcium binding protein A4
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene Alias 18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98
Molecular Weight 38.4kDa
Quantity 50μg
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Gene Symbol S100A4
Structural Form Full Length/Recombinant
Termination N-Terminal
Purity ≥90% by SDS-PAGE
Purification Method PAGE
Protein Tag IgG1 Fc Tag
Protein Family Other Proteins

S100 Calcium Binding Protein A4 (S100A4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.