Please login to your online account to display your discounted pricing

Sino Biological™ S100A13 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native human S100A13 (NP_001019381.1) (Met 1-Lys 98)

Manufacturer: life technologies  LS10991HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10991HNAE50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 11.6kDa
Structural Form Full Length/Recombinant
Expression System E. coli
Endotoxin Concentration <1 EU/μg
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Gene ID (Entrez) 6284
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Symbol S100A13
Validated Application Assay Standard
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias OTTHUMP00000034802, S100 calcium-binding protein A13, protein S100-A13
Common Name S100 calcium binding protein A13

S100 Calcium Binding Protein A13 (S100A13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.