Please login to your online account to display your discounted pricing

Sino Biological™ S100A12 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the native human S100A12 (NP_005612.1) (Met 1-Glu 92)

$90.75 - $311.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias S100A12, CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11143HNAE5

DFS Item
 
life technologies
LS11143HNAE5
5μg Each for $90.75

11143HNAE50

DFS Item
 
life technologies
LS11143HNAE50
50μg Each for $311.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias S100A12, CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6

S100 Calcium Binding Protein A12 (S100A12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human S100A12 at 2μg/mL (100μL/well) can bind human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.