Please login to your online account to display your discounted pricing

Sino Biological™ S100A12 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native human S100A12 (NP_005612.1) (Met 1-Glu 92)

Manufacturer: life technologies  LS11143HNAE5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11143HNAE5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 10.6kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias S100A12, CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6
Common Name S100 calcium binding protein A12
Gene Symbol S100A12
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

S100 Calcium Binding Protein A12 (S100A12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human S100A12 at 2μg/mL (100μL/well) can bind human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.