Please login to your online account to display your discounted pricing

Sino Biological™ S100A11 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native human S100A11 (NP_005611.1) (Met 1-Thr 105)

Manufacturer: life technologies  LS11140HNAE5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11140HNAE5

  • / Each
 

Description & Specifications

Specifications

Purification Method PAGE
Quantity 5μg
Structural Form Full Length/Recombinant
Gene Symbol S100A11
Validated Application Assay Standard
Expression System E. coli
Molecular Weight 24kDa
Purity ≥90% by SDS-PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias MLN70, S100C
Common Name S100 calcium binding protein A11
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 6282

S100 Calcium Binding Protein A11 (S100A11), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.