missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ S100A10 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100A10 (NP_002957.1) (Pro 2-Lys 97) fused to a polyhistidine tag at the N-terminus

$111.00 - $359.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, mgC111133, P11, p10
Gene ID (Entrez) 6281
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11144H07E25 Life Technologies
LS11144H07E25
5 x 5μg Each for $242.00
11144H07E5 Life Technologies
LS11144H07E5
5μg Each for $111.00
11144H07E50 Life Technologies
LS11144H07E50
50μg Each for $359.00
Specifications

Description

S100 calcium binding protein A10 (S100A10), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Frozen
6281
PAGE
ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, mgC111133, P11, p10