Please login to your online account to display your discounted pricing

Sino Biological™ S100A10 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human S100A10 (NP_002957.1) (Pro 2-Lys 97) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11144H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11144H07E5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 12.6kDa
Quantity 5μg
Format Frozen
Structural Form Full Length/Recombinant
Protein Family Other Proteins
Gene Symbol S100A10
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Common Name S100 Calcium Binding Protein A10
Gene Alias ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, mgC111133, P11, p10
Validated Application Assay Standard
Protein Tag N-His
Purification Method PAGE
Expression System E. coli
Gene ID (Entrez) 6281

S100 calcium binding protein A10 (S100A10), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.