Please login to your online account to display your discounted pricing

Sino Biological™ S100A1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human S100A1 (NP_006262.1) (Gly 2-Ser94) fused to the Fc region of human IgG1 at the N-terminus

$110.00 - $1,124.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias S100, S100-alpha, S100A
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10179H01H25

DFS Item
 
life technologies
LS10179H01H25
5 x 5μg Each for $241.00

10179H01H5

DFS Item
 
life technologies
LS10179H01H5
5μg Each for $110.00

10179H01HL

DFS Item
 
life technologies
LS10179H01H250
5 x 50μg Each for $1,124.00

10179H01H50

DFS Item
 
life technologies
LS10179H01H50
50μg Each for $372.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias S100, S100-alpha, S100A

S100 calcium binding protein A1 (S100A1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.