missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ RSPO3 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Met 1-Val 146) of human RSPO3 (Q9BXY4-1) fused to a polyhistidine tag at the C-terminus

$616.00 - $616.00

Specifications

Purity >96% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CRISTIN1, FLJ14440, PWTSR, THSD2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11185H08H1U Life Technologies
LS11185H08H1U50
50μg Each for $616.00
Specifications

Description

R-Spondin 3 (RSPO3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>96% by SDS-PAGE
<1 EU/μg
CRISTIN1, FLJ14440, PWTSR, THSD2
PAGE
Lyophilized
84870