missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RBP4 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse RBP4 (NP_035385.1) (Met 1-Leu 201) fused to a polyhistidine tag at the C-terminus

$101.00 - $1,232.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Rbp-4, Rbp4
View More Specs

Includes: Five vials of lyophilized protein

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50170M08H5 View Documents Life Technologies
LS50170M08H5
5μg Each for $101.00
 
50170M08HL View Documents Life Technologies
LS50170M08H250
5 x 50μg Each for $1,232.00
 
Description

Description

Retinol binding protein 4, plasma (RBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind all trans retinoic acid; the binding of retinoic acid results in the quenching of Trp fluorescence in RBP4; the 50% binding concentration (BC50) is >0.5μM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
Rbp-4, Rbp4

For Research Use Only. Not for use in diagnostic procedures.