missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PVRL3 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human PVRL3 (NP_056295.1) (Met 1-Asp 400) fused to the polyhistidine tag at the C-terminus

$224.00 - $1,356.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVRL3, CD113, CDw113, DKFZp566B0846, FLJ90624, PPR3, PRR3, PVRR3, nectin-3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10852H08H25 View Documents Invitrogen
LS10852H08H25
5 x 5μg Each for $224.00
 
10852H08H50 View Documents Invitrogen
LS10852H08H50
50μg Each for $395.00
 
10852H08HL View Documents Invitrogen
LS10852H08H250
5 x 50μg Each for $1,356.00
 
Description

Description

Poliovirus receptor-related 3 (PVRL3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized recombinant human PVRL3 at 1μg/mL (100μL/well) can bind biotinylated PVRL1/Nectin 1 with a linear range of 6.4-800ng/mL; 2. Immobilized recombinant human PVRL3 at 1μg/mL (100μL/well) can bind PVRL1/Nectin 1-Fc chimera with a linear range of 0.156-5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
PVRL3, CD113, CDw113, DKFZp566B0846, FLJ90624, PPR3, PRR3, PVRR3, nectin-3

For Research Use Only. Not for use in diagnostic procedures.