Please login to your online account to display your discounted pricing

Sino Biological™ PVRL2 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 360) of human PVRL2 (NP_002847.1) fused to the Fc region of human IgG1 at the C-terminus

$200.00 - $1,186.00

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVRL2, HVEB, PRR2, CD112, PVRR2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10005H02H25

DFS Item
 
life technologies
LS10005H02H25
5 x 5μg Each for $200.00

10005H02HL

DFS Item
 
life technologies
LS10005H02H250
5 x 50μg Each for $1,186.00
Description & Specifications

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVRL2, HVEB, PRR2, CD112, PVRR2

Poliovirus Receptor-Related 2 (PVRL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human PVRL2 at 20μg/mL (100μL/well) can bind biotinylated DNAM1 with a linear range of 0.078-5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.