Please login to your online account to display your discounted pricing

Sino Biological™ PVR Recombinant Rat Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of rat PVR (NP_058772.2) (Met 1-Gly 352) fused to a polyhistidine tag at the C-terminus

$143.00 - $1,776.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVR, Taa1, Tage4
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

80007R08H25

DFS Item
 
life technologies
LS80007R08H25
5 x 5μg Each for $355.00

80007R08H5

DFS Item
 
life technologies
LS80007R08H5
5μg Each for $143.00

80007R08H50

DFS Item
 
life technologies
LS80007R08H50
50μg Each for $568.00

80007R08HL

DFS Item
 
life technologies
LS80007R08H250
5 x 50μg Each for $1,776.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVR, Taa1, Tage4

Poliovirus receptor (PVR), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized rat PVR at 20μg/mL (100μL/well) can bind mouse CD226 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.