missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PVR (CD155) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 345) of mouse PVR precursor (NP_081790.1) fused to a polyhistidine tag at the C-terminus

$256.00 - $1,452.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4
Gene ID (Entrez) 52118
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50259M08H25 View Documents Life Technologies
LS50259M08H25
5 x 5μg Each for $256.00
 
50259M08HL View Documents Life Technologies
LS50259M08H250
5 x 50μg Each for $1,452.00
 
Description

Description

Poliovirus receptor (PVR/CD155), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to bind recombinant mouse CD226/DNAM-1; immobilized recombinant mouse PVR at 1μg/mL (100μL/well) can bind recombinant mouse CD226/DNAM-1 with a linear range of 0.78-100ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
52118
<1 EU/μg
3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4

For Research Use Only. Not for use in diagnostic procedures.